Proteintech
Manufacturer Code:200401AP150UL
Catalog # 50-556-585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0864 M RIP MPRIP MRIP p116Rip Rho interacting protein 3 RHOIP3 RIP3; M-RIP; Myosin phosphatase Rho-interacting protein; myosin phosphatase-Rho interacting protein; p116 Rho interacting protein; p116Rip; Rho interacting protein 3; Rho-interacting protein 3; Rho-interacting protein 3 (p116RIP) (RIP3); RIP3
Gene Aliases: 9530046C02; AA536749; AI647711; C76423; KIAA0864; M-RIP; mKIAA0864; MPRIP; MRIP; p116Rip; RHOIP3; RIP3
UniProt ID: (Human) Q6WCQ1, (Mouse) P97434, (Rat) Q9ERE6
Entrez Gene ID: (Human) 23164, (Mouse) 26936, (Rat) 116504
Molecular Function: G-protein modulator actin family cytoskeletal protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.