Invitrogen
Manufacturer Code:PA5109973
Catalog # PIPA5109973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 297-576 of human MPP7 (NP_775767.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110068J02Rik 2810038M04Rik 5430426E14Rik AI415104 Gm955 MAGUK p55 subfamily member 7 MPP7; MAGUK p55 subfamily member 7; membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7)
Gene Aliases: 1110068J02Rik; 2810038M04Rik; 5430426E14Rik; AI415104; Gm955; MPP7
UniProt ID: (Human) Q5T2T1, (Mouse) Q8BVD5
Entrez Gene ID: (Human) 143098, (Mouse) 75739
Molecular Function:
cell junction protein
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.