Proteintech
Manufacturer Code:129831AP150UL
Catalog # 50-172-8396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MPP7 Fusion Protein Ag3612 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAGUK p55 subfamily member 7; MAGUK p55 subfamily member 7 MPP7; membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7)
Gene Aliases: 1110068J02Rik; 2810038M04Rik; 5430426E14Rik; AI415104; Gm955; MPP7
UniProt ID: (Human) Q5T2T1, (Mouse) Q8BVD5, (Rat) Q5U2Y3
Entrez Gene ID: (Human) 143098, (Mouse) 75739, (Rat) 307035
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.