Invitrogen
Manufacturer Code:PA571779
Catalog # PIPA571779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 406-434 amino acids from the C-terminal region of human MPP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Discs large homolog 3; discs, large homolog 3; DLG3 MAGUK p55 subfamily member 3 discs large homolog 3 discs large homolog 3 membrane protein palmitoylated 3; MAGUK p55 subfamily member 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Protein MPP3
Gene Aliases: DLG3; MPP3
UniProt ID: (Human) Q13368
Entrez Gene ID: (Human) 4356
Molecular Function:
cell junction protein
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.