Novus Biologicals
Manufacturer Code:NBP185014
Catalog # NBP185014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SQDDPTWWQAKRVGDTNLRAGLIPSKGFQERRLSYRRAAGTLPSPQSLRKPPYDQPCDKETCDCEGYLKGHYVAGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Discs large homolog 3; Discs large homolog 3 discs large homolog 3 DLG3MAGUK p55 subfamily member 3 membrane protein palmitoylated 3 membrane protein palmitoylated 3 (MAGUK p55 subfamily member 3) Protein MPP3; discs, large homolog 3; MAGUK p55 subfamily member 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Protein MPP3
Gene Aliases: DLG3; MPP3
UniProt ID: (Human) Q13368
Entrez Gene ID: (Human) 4356
Molecular Function:
cell junction protein
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.