Proteintech
Manufacturer Code:146501AP150UL
Catalog # 50-172-9616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MPP3 Fusion Protein Ag6246 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Discs large homolog 3; Discs large homolog 3 DLG3 MAGUK p55 subfamily member 3 MPP3 Protein MPP3; discs, large homolog 3; MAGUK p55 subfamily member 3; MAGUK p55 subfamily member 3 {ECO:0000250|UniProtKB:O88910}; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Mpp3 membrane protein, palmitoylated 3; mpp3 {ECO:0000312|RGD:620015}; protein Dlgh3; Protein MPP3
Gene Aliases: 6430514B01; CSG18; DLG3; Dlgh3; Dusp3; MPP3
UniProt ID: (Human) Q13368, (Rat) O88954
Entrez Gene ID: (Human) 4356, (Rat) 114202, (Mouse) 13384
Molecular Function:
cell junction protein
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.