Proteintech
Manufacturer Code:142341AP150UL
Catalog # 50-172-9302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MPI Fusion Protein Ag5475 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mannose 6 phosphate isomerase mannose phosphate isomerase MPI Phosphohexomutase Phosphomannose isomerase PMI PMI1; mannose phosphate isomerase 1; Mannose-6-phosphate isomerase; Phosphohexomutase; Phosphomannose isomerase; phosphomannose isomerase 1; PMI
Gene Aliases: 1110002E17Rik; AI315153; CDG1B; MPI; Mpi-1; Mpi1; PMI; PMI1
UniProt ID: (Human) P34949, (Rat) Q68FX1, (Mouse) Q924M7
Entrez Gene ID: (Human) 4351, (Rat) 300741, (Mouse) 110119
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.