Invitrogen
Manufacturer Code:MA519354
Catalog # PIMA519354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G6 |
Immunogen | MPG (AAH14991 1 a.a. &bsim 90 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; ADPG; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; MDG N-methylpurine-DNA glycosylase AAG APNG CRA36.1 Mid1 N-methylpurine-DNA glycosylase MPG PIG11 PIG16 anpg 3&prime end of the Mid1 gene localized 68 kb upstream the humanzeta globin gene on 16p CRA36.1 (3-methyl-adenine DNA glycosylase) alkyladenine DNA glycosylase proliferation-inducing protein 11 proliferation-inducing protein 16; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; proliferation-inducing protein 11; proliferation-inducing protein 16
Gene Aliases: AAG; ADPG; ANPG; APNG; CRA36.1; MDG; MID1; MPG; PIG11; PIG16
UniProt ID: (Human) P29372
Entrez Gene ID: (Human) 4350
Molecular Function:
DNA binding protein
DNA glycosylase
damaged DNA-binding protein
glycosidase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.