Invitrogen
Manufacturer Code:PA576736
Catalog # PIPA576736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MPG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; 3-methyladenine DNA glycosylase; ADPG; alkylpurine-DNA-N-glycosylase; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; DNA-3-methyladenine glycosylase 3-alkyladenine DNA glycosylase 3-methyladenine DNA glycosidase ADPG N-methylpurine-DNA glycosylase AAG ANPG MID1; N-methylpurine-DNA glycocylase; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; proliferation-inducing protein 11; proliferation-inducing protein 16
Gene Aliases: 9830006D05; AAG; ADPG; AI326268; ANPG; APNG; CRA36.1; MDG; MID1; MPG; PIG11; PIG16
UniProt ID: (Human) P29372, (Mouse) Q04841
Entrez Gene ID: (Human) 4350, (Rat) 24561, (Mouse) 268395
Molecular Function:
DNA binding protein
DNA glycosylase
damaged DNA-binding protein
glycosidase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.