Novus Biologicals
Manufacturer Code:NBP297487AF405
Catalog # NB044124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MPDU1 fragment. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDGIF FLJ14836 HBeAg-binding protein 2 binding protein A Lec35 mannose-P-dolichol utilization defect 1 mannose-P-dolichol utilization defect 1 protein My008 PP3958 PQLC5 SL15HBEBP2BPA Suppressor of Lec15 and Lec35 glycosylation mutation homolog; HBeAg-binding protein 2 binding protein A; Mannose-P-dolichol utilization defect 1 protein; SL15; Suppressor of Lec15 and Lec35 glycosylation mutation homolog
Gene Aliases: CDGIF; HBEBP2BPA; Lec35; MPDU1; My008; PP3958; PQLC5; SL15
UniProt ID: (Human) O75352
Entrez Gene ID: (Human) 9526
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.