Abnova
Manufacturer Code:PAB20774
Catalog # 89-122-275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MPDU1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MPDU1.
For Research Use Only
Protein Aliases: CDGIF; FLJ14836; HBeAg-binding protein 2 binding protein A; HBEBP2BPA; Lec35; Mannose-P-dolichol utilization defect 1 protein; My008; PP3958; PQLC5; SL15; Suppressor of Lec15 and Lec35 glycosylation mutation homolog
Gene Aliases: CDGIF; HBEBP2BPA; Lec35; MPDU1; My008; PP3958; PQLC5; SL15
UniProt ID: (Human) O75352
Entrez Gene ID: (Human) 9526
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.