Proteintech
Manufacturer Code:200491AP150UL
Catalog # 50-559-256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain protein 44; brain protein 44 BRP44 Mitochondrial pyruvate carrier 2 MPC2; Mitochondrial pyruvate carrier 2; Protein 0-44
Gene Aliases: 0610006C01Rik; 2010002I07Rik; 2610205H19Rik; AA108335; BRP44; ESTM43; MPC2; RGD1563422
UniProt ID: (Human) O95563, (Rat) P38718, (Mouse) Q9D023
Entrez Gene ID: (Human) 25874, (Rat) 100359982, (Mouse) 70456
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.