Novus Biologicals
Manufacturer Code:NBP184233
Catalog # NBP184233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:DINDKSITDSTVRVIWAYHHEDAGEAGPKYHDSNRGTKSLRLLNPEKTSVLSTALPYFDLVNQDVPIPNKDTTYWCQMFKIPVFQEKHHVIKVEPV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBH-like monooxygenase protein 1; dJ248E1.1 DKFZP564G202 dopamine-oxygenase EC 1.14.17 EC 1.14.17.- Monooxygenase X monooxygenase DBH-like 1 MOXDBH-like monooxygenase protein 1 PRO5780; dopamine-oxygenase; Monooxygenase X; monooxygenase, DBH-like 1
Gene Aliases: dJ248E1.1; MOX; MOXD1; PRO5780; UNQ2493/PRO5780
UniProt ID: (Human) Q6UVY6
Entrez Gene ID: (Human) 26002
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.