Invitrogen
Manufacturer Code:PA5113910
Catalog # PIPA5113910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal region of human MOSC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mARC1; MARC1 mitochondrial amidoxime reducing component 1 Mitochondrial amidoxime-reducing component 1 moco sulfurase C-terminal domain-containing protein 1 MOCO sulphurase C-terminal domain containing 1 Molybdenum cofactor sulfurase C-terminal domain-containing protein 1 MOSC domain-containing protein 1 MOSC domain-containing protein 1 mitochondrial MOSC1; Mitochondrial amidoxime-reducing component 1; moco sulfurase C-terminal domain-containing protein 1; MOCO sulphurase C-terminal domain containing 1; Molybdenum cofactor sulfurase C-terminal domain-containing protein 1; MOSC domain-containing protein 1; MOSC domain-containing protein 1, mitochondrial
Gene Aliases: MARC1; MOSC1; MTARC1
UniProt ID: (Human) Q5VT66
Entrez Gene ID: (Human) 64757
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.