OriGene
Manufacturer Code:CF808464
Catalog # 50-166-8877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9A11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 6-77 of human MORF4L1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Eaf3 FWP006 HsT17725 MEAF3 MORFRG15 MRG15 S863-6; Esa1p-associated factor 3 homolog; MORF-related gene 15 protein; MORF-related gene on chromosome 15; Mortality factor 4-like protein 1; Protein MSL3-1; Transcription factor-like protein MRG15
Gene Aliases: Eaf3; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; MORF4L1; MORFRG15; MRG15; PP368; S863-6
UniProt ID: (Human) Q9UBU8
Entrez Gene ID: (Human) 10933
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.