Invitrogen
Manufacturer Code:PA596568
Catalog # PIPA596568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-100 of human MORF4L1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Esa1p-associated factor 3 homolog; MORF-related gene 15 protein; MORF-related gene on chromosome 15; Mortality factor 4-like protein 1; Protein MSL3-1; testis expressed gene 189; Testis-expressed gene 189 protein; Transcription factor-like protein MRG15
Gene Aliases: Eaf3; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; mKIAA4002; MORF4L1; MORFRG15; MRG15; PP368; S863-6; TEG-189; Tex189
UniProt ID: (Human) Q9UBU8, (Rat) Q6AYU1, (Mouse) P60762
Entrez Gene ID: (Human) 10933, (Rat) 300891, (Mouse) 21761
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.