OriGene
Manufacturer Code:TA800378
Catalog # 50-167-8280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-255 of human MORF4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CSR CSRB SEN SEN1; Mof4 family associated protein 1; MORF4 family-associated protein 1; mrfap1; mrfap1 {ECO:0000312|RGD:628631}; Protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; T-cell activation protein; T-cell activation protein-related protein
Gene Aliases: 9130413I22Rik; AL022827; MNCb-1039; Mrfap1; Pam14; Pgr1; RGD1306635
UniProt ID: (Mouse) Q9CQL7, (Rat) Q5M820
Entrez Gene ID: (Mouse) 67568, (Rat) 282585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.