Invitrogen
Manufacturer Code:PA566444
Catalog # PIPA566444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human MORF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CSR CSRB SEN SEN1; Esa1p-associated factor 3 homolog; MORF-related gene 15 protein; MORF-related gene on chromosome 15; Mortality factor 4-like protein 1; Protein MSL3-1; Transcription factor-like protein MRG15
Gene Aliases: Eaf3; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; MORF4L1; MORFRG15; MRG15; PP368; S863-6
UniProt ID: (Human) Q9UBU8
Entrez Gene ID: (Human) 10933
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.