Invitrogen
Manufacturer Code:PA5109970
Catalog # PIPA5109970
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 481-547 of human MON1B (NP_055755.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSRG1 HSV-1 stimulation-related gene 1 protein HSV-I stimulating-related protein KIAA0872 MON1 homolog B MON1B SAND2 SRG1 Vacuolar fusion protein MON1 homolog B; HSV-1 stimulation-related gene 1 protein; HSV-I stimulating-related protein; MON1 homolog B; MON1 secretory trafficking family member B; Vacuolar fusion protein MON1 homolog B
Gene Aliases: HSRG1; KIAA0872; MON1B; SAND2; SRG1
UniProt ID: (Human) Q7L1V2
Entrez Gene ID: (Human) 22879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.