Novus Biologicals
Manufacturer Code:NBP305063100UL
Catalog # NB095224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 481-547 of human MON1B (NP_055755.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSRG1HSV-I stimulating-related protein HSV-1 stimulation-related gene 1 protein KIAA0872vacuolar fusion protein MON1 homolog B MON1 homolog B (yeast) SAND2SRG1; HSV-1 stimulation-related gene 1 protein; HSV-I stimulating-related protein; MON1 homolog B; MON1 secretory trafficking family member B; Vacuolar fusion protein MON1 homolog B
Gene Aliases: HSRG1; KIAA0872; MON1B; SAND2; SRG1
UniProt ID: (Human) Q7L1V2
Entrez Gene ID: (Human) 22879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.