Invitrogen
Manufacturer Code:PA5103518
Catalog # PIPA5103518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MOGAT3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 3; 2-acylglycerol O-acyltransferase 3 acyl coenzyme A:monoacylglycerol acyltransferase 3 acyl-CoA:monoacylglycerol acyltransferase 3 DC7 DGAT2L2 DGAT2L7 Diacylglycerol acyltransferase 2-like protein 7 diacylglycerol O-acyltransferase candidate 7 hDC7 MGAT3 MOGAT3 monoacylglycerol O-acyltransferase 3 UNQ9383/PRO34208; acyl coenzyme A:monoacylglycerol acyltransferase 3; Acyl-CoA:monoacylglycerol acyltransferase 3; Diacylglycerol acyltransferase 2-like protein 7; Diacylglycerol O-acyltransferase candidate 7; hDC7; MGAT3; Monoacylglycerol O-acyltransferase 3
Gene Aliases: DC7; DGAT2L2; DGAT2L7; MGAT3; MOGAT3; UNQ9383/PRO34208
UniProt ID: (Human) Q86VF5
Entrez Gene ID: (Human) 346606
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.