Abnova
Manufacturer Code:H00346606A01
Catalog # 89-107-776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 59 a.a. &bsim 107 a.a) partial recombinant protein with GST tag.,MOGAT3 (NP_835470 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant MOGAT3.
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 3; acyl coenzyme A:monoacylglycerol acyltransferase 3; Acyl-CoA:monoacylglycerol acyltransferase 3; DC7; DGAT2L7; Diacylglycerol acyltransferase 2-like protein 7; Diacylglycerol O-acyltransferase candidate 7; hDC7; MGAT3; MGC119203; MGC119204; Monoacylglycerol O-acyltransferase 3
Gene Aliases: DC7; DGAT2L2; DGAT2L7; MGAT3; MOGAT3; UNQ9383/PRO34208
UniProt ID: (Human) Q86VF5
Entrez Gene ID: (Human) 346606
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.