Novus Biologicals
Manufacturer Code:NBP152078
Catalog # NBP152078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KESAAHILNRK corresponding to internal region according to NP_079374.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 2; acyl CoA:monoacylglycerol acyltransferase 2 Acyl-CoA:monoacylglycerol acyltransferase 2 DC5 DGAT2L5MGC119183 Diacylglycerol acyltransferase 2-like protein 5 Diacylglycerol O-acyltransferase candidate 5 EC 2.3.1 EC 2.3.1.22 FLJ22644 hDC5 hMGAT2 MGAT2MGC119184 MGC119185 monoacylglycerol O-acyltransferase 22-acylglycerol O-acyltransferase 2; Acyl-CoA:monoacylglycerol acyltransferase 2; Diacylglycerol acyltransferase 2-like protein 5; Diacylglycerol O-acyltransferase candidate 5; hDC5; MGAT2; Monoacylglycerol O-acyltransferase 2
Gene Aliases: DC5; DGAT2L5; DGAT2L5.; hDC5; MGAT2; MOGAT2
UniProt ID: (Human) Q3SYC2
Entrez Gene ID: (Human) 80168
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.