Abnova
Manufacturer Code:PAB2354
Catalog # 89-125-244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of mouse Mogat2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Mogat2.
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 2; acyl CoA:monoacylglycerol acyltransferase 2; Acyl-CoA:monoacylglycerol acyltransferase 2; DGAT2L5; Diacylglycerol acyltransferase 2-like protein 5; Mgat1l; MGAT2; Monoacylglycerol O-acyltransferase 1-like; Monoacylglycerol O-acyltransferase 2
Gene Aliases: DGAT2L5; Mgat1l; MGAT2; Mogat2
UniProt ID: (Mouse) Q80W94
Entrez Gene ID: (Mouse) 233549
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.