Proteintech
Manufacturer Code:195141AP150UL
Catalog # 50-173-2428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 2; acyl CoA:monoacylglycerol acyltransferase 2; Acyl-CoA:monoacylglycerol acyltransferase 2; DC5 DGAT2L5 hDC5 hMGAT2 MGAT2 MOGAT2; Diacylglycerol acyltransferase 2-like protein 5; Diacylglycerol O-acyltransferase candidate 5; hDC5; MGAT2; Monoacylglycerol O-acyltransferase 1-like; Monoacylglycerol O-acyltransferase 2
Gene Aliases: DC5; DGAT2L5; DGAT2L5.; hDC5; Mgat1l; MGAT2; MOGAT2
UniProt ID: (Human) Q3SYC2, (Mouse) Q80W94
Entrez Gene ID: (Human) 80168, (Mouse) 233549
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.