Abnova
Manufacturer Code:H00080168K
Catalog # 89-204-154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MOGAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MOGAT2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 2; Acyl-CoA:monoacylglycerol acyltransferase 2; DGAT2L5 FLJ22644 MGAT2 MGC119183 MGC119184 MGC119185; Diacylglycerol acyltransferase 2-like protein 5; Diacylglycerol O-acyltransferase candidate 5; hDC5; MGAT2; Monoacylglycerol O-acyltransferase 2
Gene Aliases: DC5; DGAT2L5; DGAT2L5.; hDC5; MGAT2; MOGAT2
UniProt ID: (Human) Q3SYC2
Entrez Gene ID: (Human) 80168
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.