Invitrogen
Manufacturer Code:PA569680
Catalog # PIPA569680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human MOGAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-acylglycerol O-acyltransferase 1; Acyl CoA:monoacylglycerol acyltransferase 1; Acyl-CoA:monoacylglycerol acyltransferase 1; Diacylglycerol acyltransferase 2-like protein 1; diacylglycerol O-acyltransferase 2 like 1; diacylglycerol O-acyltransferase 2-like 1; Diacylglycerol O-acyltransferase candidate 2; hDC2; MGAT1; Monoacylglycerol O-acyltransferase 1; putative monoacylglycerol acyltransferase 1
Gene Aliases: 0610030A14Rik; 1110064N14Rik; DC2; DGAT2L; DGAT2L1; mDC2; MGAT1; MOGAT1; WI1-2612I11.1
UniProt ID: (Bovine) Q70VZ7, (Human) Q96PD6, (Mouse) Q91ZV4
Entrez Gene ID: (Bovine) 407225, (Dog) 488548, (Pig) 100524113, (Human) 116255, (Horse) 100056612, (Mouse) 68393, (Rat) 363261
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.