Invitrogen
Manufacturer Code:PA596656
Catalog # PIPA596656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 271-460 of human MOCS3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylyltransferase and sulfurtransferase MOCS3; Adenylyltransferase MOCS3; MOCS3; Molybdenum cofactor synthesis protein 3; Molybdopterin synthase sulfurylase; Molybdopterin-synthase adenylyltransferase; Molybdopterin-synthase sulfurtransferase; MPT synthase sulfurylase; Sulfur carrier protein MOCS2A adenylyltransferase; Sulfur carrier protein MOCS2A sulfurtransferase; Sulfurtransferase MOCS3; UBA4, ubiquitin-activating enzyme E1 homolog; ubiquitin-like modifier activating enzyme 4
Gene Aliases: 1700020H17Rik; MOCS3; UBA4
UniProt ID: (Human) O95396, (Mouse) A2BDX3
Entrez Gene ID: (Human) 27304, (Mouse) 69372, (Rat) 311655
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.