Invitrogen
Manufacturer Code:PA5116230
Catalog # PIPA5116230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 14-43 amino acids from the N-terminal region of human MOCS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI415403 HAMAP-Rule:MF_03052} MCBPE MOCO1 MOCO1-A MOCO1-B MOCODB MOCS2 mocs2 {ECO:0000255 MOCS2A MOCS2B MOCS2B {ECO:0000255 molybdenum cofactor molybdenum cofactor biosynthesis protein E molybdenum cofactor synthesis 2 molybdenum cofactor synthesis protein 2 large subunit molybdenum cofactor synthesis protein 2 large subunit {ECO:0000255 Molybdenum cofactor synthesis protein 2 small subunit Molybdenum cofactor synthesis protein 2A Molybdenum cofactor synthesis protein 2B molybdenum cofactor synthesis protein 2B {ECO:0000255 molybdopterin synthase catalytic subunit molybdopterin synthase catalytic subunit {ECO:0000255 molybdopterin synthase catalytic subunit molybdopterin synthase sulfur carrier subunit molybdopterin synthase small and large subunit Molybdopterin synthase sulfur carrier subunit Molybdopterin-synthase large subunit Molybdopterin-synthase small subunit MPT synthase large subunit MPTS Sulfur carrier protein MOCS2A; MOCO1-A; MOCO1-B; MOCS2A; MOCS2B; molybdenum cofactor biosynthesis protein E; Molybdenum cofactor synthesis protein 2 large subunit; Molybdenum cofactor synthesis protein 2 small subunit; Molybdenum cofactor synthesis protein 2A; Molybdenum cofactor synthesis protein 2B; Molybdopterin synthase catalytic subunit; molybdopterin synthase small and large subunit; Molybdopterin synthase sulfur carrier subunit; Molybdopterin-synthase large subunit; Molybdopterin-synthase small subunit; MPT synthase large subunit; Sulfur carrier protein MOCS2A
Gene Aliases: MCBPE; MOCO1; MOCODB; MOCS2; MPTS
UniProt ID: (Human) Q6IAI3
Entrez Gene ID: (Human) 4338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.