Proteintech
Manufacturer Code:144161AP150UL
Catalog # 50-172-9434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MOCS2 Fusion Protein Ag5761 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCBPE MOCO1 MOCO1 A MOCS2 MOCS2A MOCS2B MPTS Sulfur carrier protein MOCS2A; MOCO1-A; MOCO1-B; mocs2 {ECO:0000255|HAMAP-Rule:MF_03052}; MOCS2A; MOCS2B; MOCS2B {ECO:0000255|HAMAP-Rule:MF_03052}; molybdenum cofactor biosynthesis protein E; Molybdenum cofactor synthesis protein 2 large subunit; molybdenum cofactor synthesis protein 2 large subunit {ECO:0000255|HAMAP-Rule:MF_03052}; Molybdenum cofactor synthesis protein 2 small subunit; Molybdenum cofactor synthesis protein 2A; Molybdenum cofactor synthesis protein 2B; molybdenum cofactor synthesis protein 2B {ECO:0000255|HAMAP-Rule:MF_03052}; Molybdopterin synthase catalytic subunit; molybdopterin synthase catalytic subunit {ECO:0000255|HAMAP-Rule:MF_03052}; molybdopterin synthase small and large subunit; Molybdopterin synthase sulfur carrier subunit; Molybdopterin-synthase large subunit; Molybdopterin-synthase small subunit; MPT synthase large subunit; Sulfur carrier protein MOCS2A
Gene Aliases: AI415403; MCBPE; MOCO1; MOCODB; MOCS2; MPTS
UniProt ID: (Human) Q6IAI3, (Mouse) Q9Z223, (Rat) Q6AY59
Entrez Gene ID: (Human) 4338, (Mouse) 17434, (Rat) 294753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.