Abnova
Manufacturer Code:H00055034M05
Catalog # 89-300-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E8 |
Immunogen | MOCOS (AAH12079.1 1 a.a. ~ 888 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ20733; HMCS; MCS; MOCOS; Molybdenum cofactor sulfurase; Molybdenum cofactor sulfurtransferase; MOS
Gene Aliases: HMCS; MCS; MOCOS; MOS
UniProt ID: (Human) Q96EN8
Entrez Gene ID: (Human) 55034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.