Invitrogen
Manufacturer Code:PA530662
Catalog # PIPA530662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 283 of Human MOCOS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HMCS MCS MOS moCo sulfurase; MCS; MOCOS; Molybdenum cofactor sulfurase; Molybdenum cofactor sulfurtransferase; MOS
Gene Aliases: HMCS; MCS; MOCOS; MOS
UniProt ID: (Human) Q96EN8
Entrez Gene ID: (Human) 55034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.