Invitrogen
Manufacturer Code:PA5114101
Catalog # PIPA5114101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MNS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW546487 meiosis specific nuclear structural 1 meiosis-specific nuclear structural 1 Meiosis-specific nuclear structural protein 1 Mns1 SPATA40 spermatogenesis associated 40; meiosis-specific nuclear structural 1; Meiosis-specific nuclear structural protein 1; spermatogenesis associated 40
Gene Aliases: MNS1; SPATA40
UniProt ID: (Human) Q8NEH6
Entrez Gene ID: (Human) 55329
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.