Proteintech
Manufacturer Code:106891AP150UL
Catalog # 50-172-6516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMS2 Fusion Protein Ag1111 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1 alpha,25-dihydroxyvitamin D3-inducible; DDVit 1; DDVit 1 DDVIT1 EDAF 1 EDPF 1 EDPF1 MMS2 MMS2 homolog UB2V2 UBE2V2 UEV 2 UEV2 Vitamin D3 inducible protein; EDAF-1; EDPF-1; enterocyte differentiation promoting factor; Enterocyte differentiation-associated factor 1; enterocyte differentiation-associated factor EDAF-1; Enterocyte differentiation-promoting factor 1; methyl methanesulfonate sensitive 2; methyl methanesulfonate sensitive 2, S. cerevisiae, homolog of; MMS2 homolog; Ubc-like protein MMS2; ubiquitin conjugating enzyme E2 variant 2; Ubiquitin-conjugating enzyme E2 variant 2; ubiquitin-conjugating enzyme E2v2; Ubiquitin-conjugating enzyme variant MMS2; Vitamin D3-inducible protein
Gene Aliases: 1110021H13Rik; 4632410D19Rik; 5730524P06Rik; 6820402M05; AI848315; C81524; DDVit-1; DDVIT1; EDAF-1; EDPF-1; EDPF1; MMS2; UBE2V2; UEV-2; UEV2
UniProt ID: (Human) Q15819, (Rat) Q7M767, (Mouse) Q8BGH6
Entrez Gene ID: (Human) 7336, (Rat) 287927, (Mouse) 70620
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.