Invitrogen
Manufacturer Code:PA579688
Catalog # PIPA579688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of rat MMP-9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 92 kDa gelatinase; 92 kDa type IV collagenase; 92-kDa type IV collagenase; 92kD gelatinase; 92kD type IV collagenase; 92kDa gelatinase; 92kDa type IV collagenase; collagenase type IVB; Gel B; Gelatinase B; GELB; matrix metalloproteinase 9; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); Matrix metalloproteinase-9; Matrix metalloproteinase-9 MMP-9 92 kDa gelatinase 92 kDa type IV collagenase Gelatinase B GELB Mmp9; MMP-9
Gene Aliases: AW743869; B/MMP9; Clg4b; MMP-9; Mmp9; pro-MMP-9
UniProt ID: (Rat) P50282, (Mouse) P41245
Entrez Gene ID: (Rat) 81687, (Mouse) 17395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.