Invitrogen
Manufacturer Code:PA513199
Catalog # PIPA513199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 644-673 amino acids from the C-terminal region of human MMP9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 67 kDa matrix metalloproteinase-9; 82 kDa matrix metalloproteinase-9; 92 kDa gelatinase; 92 kDa type IV collagenase; 92kD gelatinase; 92kD type IV collagenase; 92kDa gelatinase; 92kDa type IV collagenase; collagenase type IVB; Gel B; Gelatinase B; GELB; macrophage gelatinase; matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); matrix metalloproteinase 9; matrix metalloproteinase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase); Matrix metalloproteinase-9; Matrix metalloproteinase-9 MMP-9 92 kDa type IV collagenase 92 kDa gelatinase Gelatinase B GELB CLG4B; MMP-9; type V collagenase
Gene Aliases: AW743869; B/MMP9; CLG4B; GELB; MANDP2; MMP-9; MMP9; pro-MMP-9
UniProt ID: (Human) P14780, (Mouse) P41245
Entrez Gene ID: (Human) 4318, (Mouse) 17395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.