Proteintech
Manufacturer Code:178741AP150UL
Catalog # 50-173-1847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMP8 Fusion Protein Ag12367 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLG1 HNC Matrix metalloproteinase 8 MMP 8 MMP8 Neutrophil collagenase PMNL CL PMNL collagenase; Collagenase 2; collagenase-2; matrix metalloproteinase 8; matrix metalloproteinase 8 (neutrophil collagenase); Matrix metalloproteinase-8; MMP-8; Neutrophil collagenase; neutrophil collagenease; PMN leukocyte collagenase; PMNL collagenase; PMNL-CL
Gene Aliases: BB138268; CLG1; HNC; MMP-8; MMP8; PMNL-CL
UniProt ID: (Human) P22894, (Rat) O88766, (Mouse) O70138
Entrez Gene ID: (Human) 4317, (Rat) 63849, (Mouse) 17394
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.