Proteintech
Manufacturer Code:103742AP150UL
Catalog # 50-172-6254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMP7 Fusion Protein Ag0550 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Matrilysin; Matrilysin Matrin Matrix metalloproteinase 7 MMP 7 MMP7 MPSL1 PUMP 1 Pump 1 protease PUMP1 Uterine metalloproteinase; matrilysin, uterine; Matrin; matrix metallopeptidase 7 (matrilysin, uterine); matrix metalloproteinase 7; matrix metalloproteinase 7 (matrilysin, uterine); Matrix metalloproteinase-7; MMP-7; Pump-1 protease; uterine matrilysin; Uterine metalloproteinase
Gene Aliases: MAT; MMP-7; MMP7; MPSL1; PUMP-1; PUMP1
UniProt ID: (Human) P09237
Entrez Gene ID: (Human) 4316, (Mouse) 17393
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.