Invitrogen
Manufacturer Code:MA514210
Catalog # PIMA514210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SL-1 IID4 |
Immunogen | APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EMS-2; matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1 progelatinase) matrix metalloproteinase-3 proteoglycanase stromelysin-1 transin-1; matrix metalloproteinase 3 (stromelysin 1, progelatinase); Matrix metalloproteinase-3; MMP-3; progelatinase; proteoglycanase; SL-1; stromelysin 1; Stromelysin-1; Transin-1
Gene Aliases: CHDS6; MMP-3; MMP3; SL-1; SLN-1; SLN1; STMY; STMY1; STR-1; STR1
UniProt ID: (Human) P08254, (Mouse) P28862
Entrez Gene ID: (Human) 4314, (Mouse) 17392
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.