OriGene
Manufacturer Code:TA806979
Catalog # 50-168-1340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 246-477 of human MMP3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHDS6 MMP-3 SL-1 STMY STMY1 STR1; matrix metalloproteinase 3 (stromelysin 1, progelatinase); Matrix metalloproteinase-3; MMP-3; proteoglycanase; SL-1; Stromelysin-1; Transin-1
Gene Aliases: CHDS6; MMP-3; MMP3; SL-1; STMY; STMY1; STR1
UniProt ID: (Human) P08254
Entrez Gene ID: (Human) 4314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.