Abnova
Manufacturer Code:MAB10809
Catalog # 89-109-010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F10 |
Immunogen | Recombinant protein corresponding to human MMP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant MMP3.
For Research Use Only
Protein Aliases: CHDS6; matrix metalloproteinase 3 (stromelysin 1, progelatinase); Matrix metalloproteinase-3; MGC126102; MGC126103; MGC126104; MMP-3; proteoglycanase; SL-1; STMY; STMY1; STR1; Stromelysin-1; Transin-1
Gene Aliases: CHDS6; MMP-3; MMP3; SL-1; STMY; STMY1; STR1
UniProt ID: (Human) P08254
Entrez Gene ID: (Human) 4314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.