Proteintech
Manufacturer Code:663381IG150UL
Catalog # 50-173-6769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F5A9 |
Immunogen | MMP3 Fusion Protein Ag12359 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHDS6 Matrix metalloproteinase 3 MMP 3 MMP3 SL 1 STMY STMY1 STR1 Stromelysin 1 Transin 1; EMS-2; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); Matrix metalloproteinase-3; MMP-3; progelatinase; proteoglycanase; SL-1; stromelysin; stromelysin 1; Stromelysin-1; Transin-1
Gene Aliases: CHDS6; MMP-3; MMP10; MMP3; SL-1; SLN-1; SLN1; STMY; STMY1; STR-1; STR1
UniProt ID: (Human) P08254, (Mouse) P28862
Entrez Gene ID: (Pig) 396769, (Human) 4314, (Mouse) 17392, (Rat) 171045
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.