Novus Biologicals
Manufacturer Code:NBP15945920UL
Catalog # NBP15945920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MMP23B(matrix metallopeptidase 23B) The peptide sequence was selected from the N terminal of MMP23B. Peptide sequence ILSFPRNLLSPRETRRALAAAFRMWSDVSPFSFREVAPEQPSDLRIGFYP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24.- Femalysin matrix metallopeptidase 23B matrix metalloproteinase 22 matrix metalloproteinase 23B matrix metalloproteinase in the female reproductive tract Matrix metalloproteinase-21 Matrix metalloproteinase-22 matrix metalloproteinase-23 MIFR MIFR-1MMP22femalysin MMP21 MMP-21 MMP-22 MMP-23 MMP23A; Femalysin; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; Matrix metalloproteinase-21; Matrix metalloproteinase-22; Matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR-1; MMP-21; MMP-22; MMP-23
Gene Aliases: MIFR; MIFR-1; MMP21; MMP22; MMP23A; MMP23B
UniProt ID: (Human) O75900
Entrez Gene ID: (Human) 8510
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.