Proteintech
Manufacturer Code:130201AP150UL
Catalog # 50-172-8423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMP23B Fusion Protein Ag4113 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Femalysin; Femalysin matrix metallopeptidase 23B Matrix metalloproteinase 21 Matrix metalloproteinase 22 Matrix metalloproteinase 23 MIFR MIFR 1 MMP 21 MMP 22 MMP 23 MMP21 MMP22 MMP23A MMP23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; Matrix metalloproteinase-21; Matrix metalloproteinase-22; Matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR-1; MMP-21; MMP-22; MMP-23
Gene Aliases: MIFR; MIFR-1; MMP21; MMP22; MMP23A; MMP23B
UniProt ID: (Human) O75900
Entrez Gene ID: (Human) 8510
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.