Invitrogen
Manufacturer Code:PA125234
Catalog # PIPA125234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the hinge domain of human MMP-21. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matrix metalloproteinase 21; Matrix metalloproteinase-21; MMP-21
Gene Aliases: b2b2458Clo; b2b873Clo; HTX7; mmp; MMP-21; MMP21; mmp21-A; XELAEV_18034590mg; XMMP
UniProt ID: (Human) Q8N119, (Mouse) Q8K3F2, (Xenopus) O93470
Entrez Gene ID: (Human) 118856, (Rat) 293573, (Mouse) 214766, (Xenopus) 378572
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.