Novus Biologicals
Manufacturer Code:NBP2841650.1ML
Catalog # NB032093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of mouse MMP21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24 EC 3.4.24.- matrix metallopeptidase 21 matrix metalloproteinase 21 matrix metalloproteinase-21 MMP-21; matrix metalloproteinase 21; Matrix metalloproteinase-21; MMP-21
Gene Aliases: HTX7; MMP-21; MMP21
UniProt ID: (Human) Q8N119
Entrez Gene ID: (Human) 118856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.