Proteintech
Manufacturer Code:552891AP150UL
Catalog # 50-173-6097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matrix metallopeptidase 21 Matrix metalloproteinase 21 MMP 21 MMP21; matrix metalloproteinase 21; Matrix metalloproteinase-21; MMP-21
Gene Aliases: b2b2458Clo; b2b873Clo; HTX7; MMP-21; MMP21
UniProt ID: (Human) Q8N119, (Mouse) Q8K3F2
Entrez Gene ID: (Human) 118856, (Mouse) 214766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.