Proteintech
Manufacturer Code:554671AP150UL
Catalog # 50-173-6174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Enamel metalloproteinase; Enamel metalloproteinase Enamelysin matrix metallopeptidase 20 Matrix metalloproteinase 20 MMP 20 MMP20; Enamelysin; matrix metallopeptidase 20 (enamelysin); matrix metalloproteinase 20 (enamelysin); Matrix metalloproteinase-20; MMP-20
Gene Aliases: AI2A2; MMP-20; MMP20
UniProt ID: (Human) O60882, (Mouse) P57748
Entrez Gene ID: (Human) 9313, (Mouse) 30800, (Rat) 300341
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.