OriGene
Manufacturer Code:CF806846
Catalog # 50-166-8002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 228-507 of human MMP2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 72 kDa gelatinase; 72 kDa type IV collagenase; CLG4 CLG4A MMP-2 MMP-II MONA TBE-1; collagenase type IV-A; Gelatinase A; matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); Matrix metalloproteinase-2; matrix metalloproteinase-II; MMP-2; neutrophil gelatinase; PEX; TBE-1
Gene Aliases: CLG4; CLG4A; MMP-2; MMP-II; MMP2; MONA; TBE-1
UniProt ID: (Human) P08253
Entrez Gene ID: (Human) 4313
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.