Proteintech
Manufacturer Code:142441AP150UL
Catalog # 50-172-9312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMP19 Fusion Protein Ag5501 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matrix metallopeptidase 19 Matrix metalloproteinase 18 Matrix metalloproteinase 19 Matrix metalloproteinase RASI MMP 18 MMP 19 MMP18 MMP19 RASI RASI 1; matrix metalloproteinase 19; Matrix metalloproteinase RASI; Matrix metalloproteinase-18; Matrix metalloproteinase-19; matrix metalloproteinase-beta19; MMP-18; MMP-19
Gene Aliases: CODA; MMP18; MMP19; RASI; RASI-1
UniProt ID: (Human) Q99542, (Mouse) Q9JHI0
Entrez Gene ID: (Human) 4327, (Mouse) 58223, (Rat) 304608
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.